4-Bromo-4,4-difluoro-1-phenyl-1,3-butanedione
Dodavatel: Apollo Scientific
PC450154-1G
PC450154-250MG
APOSPC450154-1GEA
12240
CZK
APOSPC450154-1G
APOSPC450154-250MG
4-Bromo-4,4-difluoro-1-phenyl-1,3-butanedione
4-Bromo-4,4-difluoro-1-phenyl-1,3-butanedione
Vzorec:
C₁₀H₇BrF₂O₂ MW: 277,062 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18074362 CAS číslo: 1032273-54-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...