Tisk…

3-Bromo-2-fluoroprop-1-ene

Dodavatel: Apollo Scientific
Varování

PC450149-1G PC450149-250MG PC450149-5G
APOSPC450149-1GEA 8070 CZK
APOSPC450149-1G APOSPC450149-250MG APOSPC450149-5G
3-Bromo-2-fluoroprop-1-ene
3-Bromo-2-fluoroprop-1-ene
Vzorec: C₃H₄BrF
MW: 138,966 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20492668
CAS číslo: 35386-83-5
UN: 1993
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR