2,3,3,3-Tetrafluoro-1-iodopropene 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC450142-1G
PC450142-5G
APOSPC450142-1GEA
9330
CZK
APOSPC450142-1G
APOSPC450142-5G
2,3,3,3-Tetrafluoro-1-iodopropene 95%
2,3,3,3-Tetrafluoro-1-iodopropene
Vzorec:
C₃HF₄I MW: 239,938 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07784213 CAS číslo: 672339-38-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...