Tisk…

5,5,5-Trifluoropentan-2-ol

Dodavatel: Apollo Scientific
Varování

PC450130-1G PC450130-250MG PC450130-5G
APOSPC450130-1GEA 9200 CZK
APOSPC450130-1G APOSPC450130-250MG APOSPC450130-5G
5,5,5-Trifluoropentan-2-ol
5,5,5-Trifluoropentan-2-ol
Vzorec: C₅H₉F₃O
MW: 142.12 g/mol
Bod varu: 130…131 °C
Storage Temperature: Refrigerator
MDL Number: MFCD20353526
CAS číslo: 352-58-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR