2-Chloro-5-fluoro-4-thiocyanopyrimidine
Dodavatel: Apollo Scientific
PC450126-1G
PC450126-5G
APOSPC450126-1GEA
5660
CZK
APOSPC450126-1G
APOSPC450126-5G
2-Chloro-5-fluoro-4-thiocyanopyrimidine
2-Chloro-5-fluoro-4-thiocyanopyrimidine
Vzorec:
C₅HClFN₃S MW: 189.598 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18072176 CAS číslo: 112889-53-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...