Tisk…

2-(Chlorodifluoromethyl)-1,3-benzoxazole

Dodavatel: Apollo Scientific

PC450125-1G PC450125-5G
APOSPC450125-1GEA 5200 CZK
APOSPC450125-1G APOSPC450125-5G
2-(Chlorodifluoromethyl)-1,3-benzoxazole
2-(Chlorodifluoromethyl)-1,3-benzoxazole
Vzorec: C₈H₄ClF₂NO
MW: 203,573 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18074399
CAS číslo: 212311-54-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR