Tisk…

3-(Chlorodifluoromethyl)-5-methyl-1H-pyrazole

Dodavatel: Apollo Scientific
Varování

PC450117-1G PC450117-5G
APOSPC450117-1GEA 910 CZK
APOSPC450117-1G APOSPC450117-5G
3-(Chlorodifluoromethyl)-5-methyl-1H-pyrazole
3-(Chlorodifluoromethyl)-5-methyl-1H-pyrazole
Vzorec: C₅H₅ClF₂N₂
MW: 166,556 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04969954
CAS číslo: 321998-14-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR