Tisk…

Bis(2,2,3,3,3-pentafluoropropyl)ether

Dodavatel: Apollo Scientific

PC450115-1G PC450115-5G
APOSPC450115-1GEA 5390 CZK
APOSPC450115-1G APOSPC450115-5G
Bis(2,2,3,3,3-pentafluoropropyl)ether
Bis(2,2,3,3,3-pentafluoropropyl)ether
Vzorec: C₆H₄F₁₀O
MW: 282.079 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22123959
CAS číslo: 1691-22-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR