Tisk…

(1,1-Difluoroethyl)benzene

Dodavatel: Apollo Scientific
Varování

PC450090-1G PC450090-5G
APOSPC450090-1GEA 8730 CZK
APOSPC450090-1G APOSPC450090-5G
(1,1-Difluoroethyl)benzene
(1,1-Difluoroethyl)benzene
Vzorec: C₈H₈F₂
MW: 142,146 g/mol
Bod varu: 67 °C (47 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD11975315
CAS číslo: 657-35-2
UN: 1993
ADR: 3,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR