Tisk…

3-(1H,1H-Heptafluorobutoxy)propane-1,2-diol 95%

Dodavatel: Apollo Scientific
Varování

PC450087-1G
APOSPC450087-1GEA 0 CZK
APOSPC450087-1G
3-(1H,1H-Heptafluorobutoxy)propane-1,2-diol 95%
3-(1H,1H-Heptafluorobutoxy)propane-1,2-diol
Vzorec: C₇H₉F₇O₃
MW: 274,133 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22123958
CAS číslo: 25385-72-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR