3-(Bromodifluoromethyl)-5-phenyl-1H-pyrazole
Dodavatel: Apollo Scientific
PC450074-1G
PC450074-5G
APOSPC450074-1GEA
8100
CZK
APOSPC450074-1G
APOSPC450074-5G
3-(Bromodifluoromethyl)-5-phenyl-1H-pyrazole
3-(Bromodifluoromethyl)-5-phenyl-1H-pyrazole
Vzorec:
C₁₀H₇BrF₂N₂ MW: 273,077 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD19442114 CAS číslo: 1355046-54-6 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...