Tisk…

2-Chloro-2,2-difluoro-N-(2'-hydroxyphenyl)acetamide

Dodavatel: Apollo Scientific

PC450073-1G PC450073-5G
APOSPC450073-1GEA 3540 CZK
APOSPC450073-1G APOSPC450073-5G
2-Chloro-2,2-difluoro-N-(2'-hydroxyphenyl)acetamide
2-Chloro-2,2-difluoro-N-(2'-hydroxyphenyl)acetamide
Vzorec: C₈H₆ClF₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD18072172
CAS číslo: 1309602-47-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR