Potassium trifluoro(pentafluoroethyl)borate
Dodavatel: Apollo Scientific
|
Varování
|
PC450058-1G
PC450058-5G
APOSPC450058-1GEA
5490
CZK
APOSPC450058-1G
APOSPC450058-5G
Potassium trifluoro(pentafluoroethyl)borate
Potassium trifluoro(pentafluoroethyl)borate
Vzorec:
C₂BF₈·K MW: 225.918 g/mol Bod tání: 287 °C Storage Temperature: Ambient |
MDL Number:
MFCD17214187 CAS číslo: 476639-90-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...