Tisk…

1-(4-Chlorophenyl)-1-fluoroethene

Dodavatel: Apollo Scientific

PC450053-100MG PC450053-1G PC450053-250MG
APOSPC450053-100MGEA 2280 CZK
APOSPC450053-100MG APOSPC450053-1G APOSPC450053-250MG
1-(4-Chlorophenyl)-1-fluoroethene
1-(4-Chlorophenyl)-1-fluoroethene
Vzorec: C₈H₆ClF
MW: 156,585 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22123956
CAS číslo: 106263-12-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR