Tisk…

1-(2-Fluoroethyl)piperazine dihydrochloride 98%

Dodavatel: Apollo Scientific
Varování

PC449008-1G PC449008-5G
APOSPC449008-1GEA 1580 CZK
APOSPC449008-1G APOSPC449008-5G
1-(2-Fluoroethyl)piperazine dihydrochloride 98%
1-(2-Fluoroethyl)piperazine dihydrochloride
Vzorec: C₆H₁₃FN₂·2ClH
MW: 205.101 g/mol
Storage Temperature: Ambient
MDL Number: MFCD21602355
CAS číslo: 1089279-64-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR