2-(4-Fluorophenyl)amino-4-(2-hydroxyphenyl)-1,3-thiazole hydrobromide 97%
Dodavatel: Apollo Scientific
|
Varování
|
PC4484-1G
PC4484-5G
APOSPC4484-1GEA
2980
CZK
APOSPC4484-1G
APOSPC4484-5G
2-(4-Fluorophenyl)amino-4-(2-hydroxyphenyl)-1,3-thiazole hydrobromide 97%
2-(4-Fluorophenyl)amino-4-(2-hydroxyphenyl)-1,3-thiazole hydrobromide
Vzorec:
C₁₅H₁₁FN₂OS·BrH Bod tání: 205…210 °C Storage Temperature: Ambient |
MDL Number:
MFCD08458094 CAS číslo: 1211182-74-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...