tert-Butyl 2-amino-4-(trifluoromethyl)benzoate
Dodavatel: Apollo Scientific
PC448058-1G
PC448058-250MG
APOSPC448058-1GEA
27970
CZK
APOSPC448058-1G
APOSPC448058-250MG
tert-Butyl 2-amino-4-(trifluoromethyl)benzoate
tert-Butyl 2-amino-4-(trifluoromethyl)benzoate
Vzorec:
C₁₂H₁₄F₃NO₂ MW: 261.24 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD08274733 CAS číslo: 1485729-20-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...