5-(Trifluoromethoxy)pyridin-2-amine 96%
Dodavatel: Apollo Scientific
|
Varování
|
PC448054-1G
PC448054-100MG
PC448054-250MG
APOSPC448054-1GEA
28110
CZK
APOSPC448054-1G
APOSPC448054-100MG
APOSPC448054-250MG
5-(Trifluoromethoxy)pyridin-2-amine 96%
5-(Trifluoromethoxy)pyridin-2-amine
Vzorec:
C₆H₅F₃N₂O Storage Temperature: Ambient |
MDL Number:
MFCD19690149 CAS číslo: 1221171-88-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...