Tisk…

3-Fluoro-4-(hydroxymethyl)benzeneboronic acid 98%

Dodavatel: Apollo Scientific
Varování

PC448015-250MG PC448015-1G PC448015-5G
APOSPC448015-250MGEA 1040 CZK
APOSPC448015-250MG APOSPC448015-1G APOSPC448015-5G
3-Fluoro-4-(hydroxymethyl)benzeneboronic acid 98%
3-Fluoro-4-(hydroxymethyl)benzeneboronic acid
Vzorec: C₇H₈BFO₃
Storage Temperature: Ambient
MDL Number: MFCD13181609
CAS číslo: 1082066-52-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR