Triphenylsulphonium nonaflate 99%
Dodavatel: Apollo Scientific
|
Varování
|
PC4472-1G
PC4472-250MG
PC4472-5G
APOSPC4472-1GEA
9660
CZK
APOSPC4472-1G
APOSPC4472-250MG
APOSPC4472-5G
Triphenylsulphonium nonaflate 99%
Triphenylsulphonium nonaflate
Photoinitiator/photoacid generator
Vzorec:
C₁₈H₁₅S·C₄F₉O₃S MW: 562,468 g/mol Bod tání: 84…88 °C Storage Temperature: Ambient |
MDL Number:
MFCD02683476 CAS číslo: 144317-44-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...