Tisk…

4-Chloro-3-fluorobenzenesulphonyl chloride 95+%

Dodavatel: Apollo Scientific

PC446151-1G PC446151-5G
APOSPC446151-1GEA 3270 CZK
APOSPC446151-1G APOSPC446151-5G
4-Chloro-3-fluorobenzenesulphonyl chloride 95+%
4-Chloro-3-fluorobenzenesulphonyl chloride
Vzorec: C₆H₃Cl₂FO₂S
MW: 229,056 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09859891
CAS číslo: 942035-77-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR