5-Chloro-3-(3-fluorophenyl)-1,2,4-thiadiazole 95+%
Dodavatel: Apollo Scientific
PC446089-1G
PC446089-5G
APOSPC446089-1GEA
5460
CZK
APOSPC446089-1G
APOSPC446089-5G
5-Chloro-3-(3-fluorophenyl)-1,2,4-thiadiazole 95+%
5-Chloro-3-(3-fluorophenyl)-1,2,4-thiadiazole
Vzorec:
C₈H₄ClFN₂S MW: 214,647 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11052448 CAS číslo: 138426-26-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...