Tisk…

3-Cyano-5-methyl-2-(trifluoromethyl)furan 95+%

Dodavatel: Apollo Scientific

PC446086-1G PC446086-5G
APOSPC446086-1GEA 3270 CZK
APOSPC446086-1G APOSPC446086-5G
3-Cyano-5-methyl-2-(trifluoromethyl)furan 95+%
3-Cyano-5-methyl-2-(trifluoromethyl)furan
Vzorec: C₇H₄F₃NO
MW: 175,108 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00525589
CAS číslo: 1053656-28-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR