2-Phenyl-5-trifluoromethyl-[1,3,4]oxadiazole
Dodavatel: Apollo Scientific
PC446060-1G
PC446060-500MG
APOSPC446060-1GEA
9030
CZK
APOSPC446060-1G
APOSPC446060-500MG
2-Phenyl-5-trifluoromethyl-[1,3,4]oxadiazole
2-Phenyl-5-trifluoromethyl-[1,3,4]oxadiazole
Vzorec:
C₉H₅F₃N₂O MW: 214,144 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00488311 CAS číslo: 298207-18-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...