Tisk…

2,6-Bis-trifluoromethyl-nicotinic acid ethyl ester

Dodavatel: Apollo Scientific

PC446059-1G PC446059-5G
APOSPC446059-1GEA 5720 CZK
APOSPC446059-1G APOSPC446059-5G
2,6-Bis-trifluoromethyl-nicotinic acid ethyl ester
2,6-Bis-trifluoromethyl-nicotinic acid ethyl ester
Vzorec: C₁₀H₇F₆NO₂
MW: 287,158 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13190031
CAS číslo: 1095211-28-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR