6-Fluoro-quinoxaline-2,3-diol
Dodavatel: Apollo Scientific
PC446058-1G
PC446058-500MG
APOSPC446058-1GEA
9030
CZK
APOSPC446058-1G
APOSPC446058-500MG
6-Fluoro-quinoxaline-2,3-diol
6-Fluoro-quinoxaline-2,3-diol
Vzorec:
C₈H₅FN₂O₂ MW: 180,136 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00629759 CAS číslo: 61875-34-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...