2-Chloromethyl-5-trifluoromethyl-benzothiazole 90%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
PC446043-100MG
PC446043-250MG
APOSPC446043-100MGEA
5000
CZK
APOSPC446043-100MG
APOSPC446043-250MG
2-Chloromethyl-5-trifluoromethyl-benzothiazole 90%
2-Chloromethyl-5-trifluoromethyl-benzothiazole
Vzorec:
C₉H₅ClF₃NS MW: 251,656 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11040746 CAS číslo: 110704-50-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...