Tisk…

(6-Chloro-4-trifluoromethyl-pyridin-2-yloxy)-acetic acid methyl ester

Dodavatel: Apollo Scientific

PC446038-1G PC446038-5G
APOSPC446038-1GEA 5720 CZK
APOSPC446038-1G APOSPC446038-5G
(6-Chloro-4-trifluoromethyl-pyridin-2-yloxy)-acetic acid methyl ester
(6-Chloro-4-trifluoromethyl-pyridin-2-yloxy)-acetic acid methyl ester
Vzorec: C₉H₇ClF₃NO₃
MW: 269,605 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11845743
CAS číslo: 1160994-91-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR