2-Bromomethyl-6-chloro-4-trifluoromethyl-pyridine
Dodavatel: Apollo Scientific
PC446036-1G
PC446036-500MG
APOSPC446036-1GEA
14750
CZK
APOSPC446036-1G
APOSPC446036-500MG
2-Bromomethyl-6-chloro-4-trifluoromethyl-pyridine
2-Bromomethyl-6-chloro-4-trifluoromethyl-pyridine
Vzorec:
C₇H₄BrClF₃N MW: 274,466 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11227256 CAS číslo: 862120-75-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...