Tisk…

Ethyl 2-(4-(trifluoromethy)phenoxy)acetoacetate

Dodavatel: Apollo Scientific

PC446025-1G PC446025-5G
APOSPC446025-1GEA 5200 CZK
APOSPC446025-1G APOSPC446025-5G
Ethyl 2-(4-(trifluoromethy)phenoxy)acetoacetate
Ethyl 2-(4-(trifluoromethy)phenoxy)acetoacetate
Vzorec: C₁₃H₁₃F₃O₄
Storage Temperature: Ambient
MDL Number: MFCD10568347
CAS číslo: 1053657-44-5

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR