Tisk…

5-Chloromethyl-2-(4-(trifluoromethyl)phenyl)-1H-imidazole

Dodavatel: Apollo Scientific

PC446024-1G PC446024-5G
APOSPC446024-1GEA 5200 CZK
APOSPC446024-1G APOSPC446024-5G
5-Chloromethyl-2-(4-(trifluoromethyl)phenyl)-1H-imidazole
5-Chloromethyl-2-(4-(trifluoromethyl)phenyl)-1H-imidazole
Vzorec: C₁₁H₈ClF₃N₂
MW: 260,643 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10568333
CAS číslo: 466664-94-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR