Tisk…

5-(tert-Butyloxycarbonyamino)methyl-3-(4-fluorophenyl)-[1,2,4]oxadiazole

Dodavatel: Apollo Scientific

PC446023-1G PC446023-5G
APOSPC446023-1GEA 5200 CZK
APOSPC446023-1G APOSPC446023-5G
5-(tert-Butyloxycarbonyamino)methyl-3-(4-fluorophenyl)-[1,2,4]oxadiazole
5-(tert-Butyloxycarbonyamino)methyl-3-(4-fluorophenyl)-[1,2,4]oxadiazole
Vzorec: C₁₄H₁₆FN₃O₃
MW: 293,294 g/mol
Storage Temperature: Ambient
MDL Number: MFCD10568311
CAS číslo: 1053656-53-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR