2-(Trifluoromethoxy)benzamidoxime
Dodavatel: Apollo Scientific
|
Varování
|
PC446017-10G
PC446017-1G
APOSPC446017-10GEA
11150
CZK
APOSPC446017-10G
APOSPC446017-1G
2-(Trifluoromethoxy)benzamidoxime
2-(Trifluoromethoxy)benzamidoxime
Vzorec:
C₈H₇F₃N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD07772878 CAS číslo: 63968-84-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...