Ammonium 2-perfluoropentoxy-2,3,3,3-tetrafluoropropanoate
Dodavatel: Apollo Scientific
|
Varování
|
PC4451-100G
PC4451-25G
PC4451-5G
APOSPC4451-100GEA
29590
CZK
APOSPC4451-100G
APOSPC4451-25G
APOSPC4451-5G
Ammonium 2-perfluoropentoxy-2,3,3,3-tetrafluoropropanoate
Ammonium 2-perfluoropentoxy-2,3,3,3-tetrafluoropropanoate
Vzorec:
C₈HF₁₅O₃·H₃N Storage Temperature: Ambient |
MDL Number:
MFCD07783987 CAS číslo: 96513-97-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...