Tisk…

4-Amino-3-fluorophenylacetic acid 97%

Dodavatel: Apollo Scientific

PC4414-100MG PC4414-250MG PC4414-1G
APOSPC4414-100MGEA 3110 CZK
APOSPC4414-100MG APOSPC4414-250MG APOSPC4414-1G
4-Amino-3-fluorophenylacetic acid 97%
4-Amino-3-fluorophenylacetic acid
Vzorec: C₈H₈FNO₂
MW: 169,153 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094244
CAS číslo: 503315-77-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR