Methyl 2-chloro-3,5-difluorobenzoate 95%
Dodavatel: Apollo Scientific
|
Varování
|
PC43512-1G
PC43512-250MG
PC43512-5G
APOSPC43512-1GEA
1660
CZK
APOSPC43512-1G
APOSPC43512-250MG
APOSPC43512-5G
Methyl 2-chloro-3,5-difluorobenzoate 95%
Methyl 2-chloro-3,5-difluorobenzoate
Vzorec:
C₈H₅ClF₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD18417405 CAS číslo: 1261846-02-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...