Tisk…

Perfluoro(ethylcyclohexane) 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: Undecafluoro(pentafluoroethyl)cyclohexane

PC4325-25G PC4325-5G
APOSPC4325-25GEA 4340 CZK
APOSPC4325-25G APOSPC4325-5G
Perfluoro(ethylcyclohexane) 95%
Perfluoro(ethylcyclohexane)
Vzorec: C₈F₁₆
Bod varu: 100…102 °C
Bod tání: –60 °C
Storage Temperature: Ambient
MDL Number: MFCD00066613
CAS číslo: 335-21-7

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR