1-(tert-Butoxycarbonyl)-5-fluoropiperidine-3-carboxylic acid 95+%
Dodavatel: Apollo Scientific
|
Varování
|
PC430287-100MG
PC430287-250MG
APOSPC430287-100MGEA
5100
CZK
APOSPC430287-100MG
APOSPC430287-250MG
1-(tert-Butoxycarbonyl)-5-fluoropiperidine-3-carboxylic acid 95+%
1-(tert-Butoxycarbonyl)-5-fluoropiperidine-3-carboxylic acid
Vzorec:
C₁₁H₁₈FNO₄ Storage Temperature: Ambient |
MDL Number:
MFCD17015060 CAS číslo: 1241725-64-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...