Tisk…

Methyl 2-fluoro-3-methoxybenzoate 98%

Dodavatel: Apollo Scientific

PC430003-5G
APOSPC430003-5GEA 3970 CZK
APOSPC430003-5G
Methyl 2-fluoro-3-methoxybenzoate 98%
Methyl 2-fluoro-3-methoxybenzoate
Vzorec: C₉H₉FO₃
MW: 184,164 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14698048
CAS číslo: 958991-48-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR