3-Amino-2-bromo-6-fluoropyridine 97%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
2-Bromo-6-fluoropyridin-3-amine
PC42590-1G
PC42590-5G
PC42590-10G
APOSPC42590-1GEA
622
CZK
APOSPC42590-1G
APOSPC42590-5G
APOSPC42590-10G
3-Amino-2-bromo-6-fluoropyridine 97%
3-Amino-2-bromo-6-fluoropyridine
Vzorec:
C₅H₄BrFN₂ MW: 191,001 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD18262185 CAS číslo: 1068976-51-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...