8-Chloro-3-fluoro-1,5-naphthyridine 95%
Dodavatel: Apollo Scientific
PC421228-100MG
PC421228-1G
PC421228-250MG
APOSPC421228-100MGEA
9260
CZK
APOSPC421228-100MG
APOSPC421228-1G
APOSPC421228-250MG
8-Chloro-3-fluoro-1,5-naphthyridine 95%
8-Chloro-3-fluoro-1,5-naphthyridine
Vzorec:
C₈H₄ClFN₂ MW: 182,582 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD28098174 CAS číslo: 1151802-02-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...