5,8-Difluoro-2H-chromene 97%
Dodavatel: Apollo Scientific
PC421221-100MG
PC421221-1G
PC421221-250MG
APOSPC421221-100MGEA
12200
CZK
APOSPC421221-100MG
APOSPC421221-1G
APOSPC421221-250MG
5,8-Difluoro-2H-chromene 97%
5,8-Difluoro-2H-chromene
Vzorec:
C₉H₆F₂O Storage Temperature: Ambient |
MDL Number:
MFCD09909897 CAS číslo: 457628-37-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...