Tisk…

4-Cyclopropyl-2-fluoro-6-methylbenzamide 95%

Dodavatel: Apollo Scientific

PC421217-100MG PC421217-1G PC421217-250MG
APOSPC421217-100MGEA 12800 CZK
APOSPC421217-100MG APOSPC421217-1G APOSPC421217-250MG
4-Cyclopropyl-2-fluoro-6-methylbenzamide 95%
4-Cyclopropyl-2-fluoro-6-methylbenzamide
Vzorec: C₁₁H₁₂FNO
MW: 193,217 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20726776
CAS číslo: 1242156-52-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR