Tisk…

4-Bromo-2-fluoro-6-methylbenzamide 97%

Dodavatel: Apollo Scientific

PC421214-100MG PC421214-250MG
APOSPC421214-100MGEA 2440 CZK
APOSPC421214-100MG APOSPC421214-250MG
4-Bromo-2-fluoro-6-methylbenzamide 97%
4-Bromo-2-fluoro-6-methylbenzamide
Vzorec: C₈H₇BrFNO
MW: 232,05 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28405379
CAS číslo: 1242156-51-9

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR