Tisk…

2-(4-Fluoronaphthalen-1-yl)acetonitrile 98%

Dodavatel: Apollo Scientific

PC421201-100MG PC421201-1G PC421201-250MG
APOSPC421201-100MGEA 6390 CZK
APOSPC421201-100MG APOSPC421201-1G APOSPC421201-250MG
2-(4-Fluoronaphthalen-1-yl)acetonitrile 98%
2-(4-Fluoronaphthalen-1-yl)acetonitrile
Vzorec: C₁₂H₈FN
Storage Temperature: Ambient
MDL Number: MFCD09923719
CAS číslo: 3832-87-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR