Tisk…

(E)-4-(2-Fluorophenyl)but-3-enoic acid 98%

Dodavatel: Apollo Scientific

PC421190-100MG PC421190-1G PC421190-250MG
APOSPC421190-100MGEA 10250 CZK
APOSPC421190-100MG APOSPC421190-1G APOSPC421190-250MG
(E)-4-(2-Fluorophenyl)but-3-enoic acid 98%
(E)-4-(2-Fluorophenyl)but-3-enoic acid
Vzorec: C₁₀H₉FO₂
MW: 180,176 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18324916
CAS číslo: 127406-53-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR