1-(Tetrahydro-2h-pyran-2-yl)-5-(trifluoromethyl)-1h-indazol-4-ylboronic acid
Dodavatel: Apollo Scientific
Synonyms:
(1-(Tetrahydro-2H-pyran-2-yl)-5-(trifluoromethyl)-1H-indazol-4-yl)boronic acid
PC421179-100MG
PC421179-250MG
APOSPC421179-100MGEA
1740
CZK
APOSPC421179-100MG
APOSPC421179-250MG
1-(Tetrahydro-2h-pyran-2-yl)-5-(trifluoromethyl)-1h-indazol-4-ylboronic acid
1-(Tetrahydro-2h-pyran-2-yl)-5-(trifluoromethyl)-1h-indazol-4-ylboronic acid
Vzorec:
C₁₃H₁₄BF₃N₂O₃ MW: 314,068 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD30609549 CAS číslo: 2374153-25-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...