Tisk…

4-Bromo-1-(difluoromethyl)-1H-pyrazole

Dodavatel: Apollo Scientific
Varování

PC421172-1G PC421172-5G PC421172-250MG
APOSPC421172-1GEA 5630 CZK
APOSPC421172-1G APOSPC421172-5G APOSPC421172-250MG
4-Bromo-1-(difluoromethyl)-1H-pyrazole
4-Bromo-1-(difluoromethyl)-1H-pyrazole
Vzorec: C₄H₃BrF₂N₂
MW: 196,981 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11558132
CAS číslo: 956477-67-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR