Tisk…

2-Chloro-6-(trifluoromethyl)isonicotinic acid 97%

Dodavatel: Apollo Scientific

PC421158-1G PC421158-5G
APOSPC421158-1GEA 2730 CZK
APOSPC421158-1G APOSPC421158-5G
2-Chloro-6-(trifluoromethyl)isonicotinic acid 97%
2-Chloro-6-(trifluoromethyl)isonicotinic acid
Vzorec: C₇H₃ClF₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD13188811
CAS číslo: 796090-23-8

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR