Tisk…

tert-Butyl 3,5-difluoro-4-formylbenzoate

Dodavatel: Apollo Scientific

PC421142-1G PC421142-250MG
APOSPC421142-1GEA 9330 CZK
APOSPC421142-1G APOSPC421142-250MG
tert-Butyl 3,5-difluoro-4-formylbenzoate
tert-Butyl 3,5-difluoro-4-formylbenzoate
Vzorec: C₁₂H₁₂F₂O₃
MW: 242.219 g/mol
MDL Number: MFCD11975649
CAS číslo: 467442-12-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR